ARISTAR®VWR® , Gallium Standard Solution, 1000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455382EEA 236 EUR
455382E 455384G
ARISTAR®VWR® , Gallium Standard Solution, 1000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Standaarden Kationische standaarden Gallium standaarden
Ga in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.