VWR® ARISTAR®, Iron Standard Solution, 1000 mg/L Fe in dil. Nitric Acid (from Fe), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455522VEA 160 EUR
455522V 455524A
VWR® ARISTAR®, Iron Standard Solution, 1000 mg/L Fe in dil. Nitric Acid (from Fe), Standard for ICP
Standaarden Kationische standaarden IJzer standaarden
Fe in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.