VWR® ARISTAR®, Lead Standard Solution, 10000 mg/L Pb in dil. Nitric Acid (from Pb(NO₃)₂), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455572YEA 311 EUR
455572Y 455574K
VWR® ARISTAR®, Lead Standard Solution, 10000 mg/L Pb in dil. Nitric Acid (from Pb(NO₃)₂), Standard for ICP
Standaarden Kationische standaarden Lood standaarden
Pb(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
Specificaties
Environmentally Preferable