VWR® ARISTAR®, Palladium Standard Solution, 1000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455762MEA 331 EUR
455762M 455764X
VWR® ARISTAR®, Palladium Standard Solution, 1000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
Standaarden Kationische standaarden Palladium standaarden
Pd in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.