Kiezelgel, Pore size: 120 Å ≥99.99%, spherical 3 µm APS for HPLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

45803.03 45803.06
45803.03EA 62.6 EUR
45803.03 45803.06
Kiezelgel, Pore size: 120 Å ≥99.99%, spherical 3 µm APS for HPLC
Kiezelgel

Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 340m2/g, P.V. 1.00cc/g MDL No.: MFCD00011232

Formule: SiO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR