Smeltkroezen
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73056 73058 73060 73061 73063 73065 73067
459-0237EA 72.3 EUR
459-0237 216-4479 216-4481 459-0176 459-0172 459-0178 459-0173
Smeltkroezen
Kroezen
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Bestelinformatie: Crucible and lid have to be ordered separately.