Verwarmingscirculatoren
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2003.0001.01 2005.0001.01 2006.0001.01 2007.0001.01 2004.0001.01 2046.0001.01 2047.0001.01
461-6456EA 2670 EUR
461-6456 461-0137 461-0138 461-0139 461-6457 HUBE2046.0001.01 HUBE2047.0001.01
Verwarmingscirculatoren
Circulatiethermostaten Open circulatiesystemen Baden met ingebouwde circulatiethermostaat
The low bath volume means these circulators are ideal for controlling the temperature of small external applications, or for direct temperature control of small objects directly in the bath.
The Pilot ONE® controller offers a brilliant 14,5 cm touch screen with graphic functions and a comfortable navigation menu in 11 languages plus USB and Ethernet connections.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • Controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade®
More Product Information
Order Now
Specificaties
Environmentally Preferable