Dexamethasone-21-acetate, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dion, Dexamethasone acetate

M-Clarity™ quality level = MQ100

46166-100MG
46166-100MGEA 84.3 EUR
46166-100MG
Dexamethasone-21-acetate, Supelco®
Dexamethasone-21-acetate
Formule: C₂₄H₃₁FO₆
MDL Nummer: MFCD00027407
CAS nummer: 1177-87-3
EINECS: 214-646-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dce1c226edc5df3adf6f33280a0b29e79fec5b7&vid=fe3111abb408bd4b0f61239ccd0be4c874daa6d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR