Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

46661.G1 46661.G5 46661.G9
46661.G1EA 128 EUR
46661.G1 46661.G5 46661.G9
Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Indium
Formule: In
MW: 114.82 g/mol
Kookpunt: 2000
Smeltpunt: 156
Dichtheid: 7.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR