Kaliumhydroxide concentrate for dilution to 1 mol/l (1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

4674.EA 43.11 EUR
4674.
Kaliumhydroxide concentrate for dilution to 1 mol/l (1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Kaliumhydroxide

(1 equiv.=56.11 g KOH)

Formule: KOH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.998-1.002
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 mL)
Low in Carbonate

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR