Tedizolid

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

467580010 467582500
467580010.EA 206 EUR
467580010. 467582500.
Tedizolid
Tedizolid
Formule: C₁₇H₁₅FN₆O₃
Smeltpunt: 201°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD19442562
CAS nummer: 856866-72-3

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 6%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR