Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H420530000
468-4101EA 640 EUR
468-4101
Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Exsiccatoren Exsiccatorkasten
Small, clear polystyrene desiccator cabinet, these can be safely stacked up to five units high for maximum use of dry storage space.
Leveringsinformatie: Supplied with ready to use jar of silica gel.
Order Now
Specificaties
Environmentally Preferable