Tranilast

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

468610010 468610050
468610010.EA 64.2 EUR
468610010. 468610050.
Tranilast
Tranilast
Formule: C₁₈H₁₇NO₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00864787
CAS nummer: 53902-12-8
Merck Index: 13,09646

Order Now

Specification Test Results

Appearance white to light yellow to green crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 207°C to 210°C
Titration with TBAH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR