Etretinate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

469960010 469962500
469960010.EA 280 EUR
469960010. 469962500.
Etretinate 98%
Etretinate
Formule: C₂₃H₃₀O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00866624
CAS nummer: 54350-48-0
EINECS: 259-119-3
UN: 3077
ADR: 9,III
Merck Index: 13,03922

Order Now

Specification Test Results

Appearance white to yellow to green crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 103°C to 107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR