N-Acetyltryptamine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

470261000 470265000
470261000.EA 109 EUR
470261000. 470265000.
N-Acetyltryptamine 98%
N-Acetyltryptamine
Formule: C₁₂H₁₄N₂O
Kookpunt: 484,7°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00209910
CAS nummer: 1016-47-3

Order Now

Specification Test Results

Appearance white to light yellow to light orange crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 74°C to 78°C
Water ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR