2-Propylpyridine-4-carbothioamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Propyl-4-pyridinecarbothioamide, 2-Propylthioisonicotinamide, 2-Propylpyridine-4-carbothioamide, Prothionamide

471200050 471200250 471201000
471200050.EA 49.3 EUR
471200050. 471200250. 471201000.
2-Propylpyridine-4-carbothioamide 98%
2-Propylpyridine-4-carbothioamide
Formule: C₉H₁₂N₂S
MW: 180.27 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00464119
CAS nummer: 14222-60-7
EINECS: 238-093-7
Merck Index: 13,07982
New Product

Order Now

Specification Test Results

Appearance yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR