Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

47152.AE 47152.AP
47152.AEEA 139 EUR
47152.AE 47152.AP
Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®
Standaarden Kationische standaarden Fosfor standaarden

Molecular Formula: H₃PO₄, 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00133771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR