VALEGRO Recirculating Coolers
Leverancier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9610050 9610035
473-3201EA 3970 EUR
473-3201 JULA9610035
VALEGRO Recirculating Coolers
Circulatiethermostaten Gesloten circulatiesystemen Koelingscirculatoren
VALEGRO recirculating coolers are environmentally-friendly all-rounders for a wide range of temperature control tasks. They are characterised by a good ratio between cooling capacity and unit size as well as optimal usability. They work with natural refrigerant and can be adapted to application requirements thanks to their adjustable pump.
  • Climate-friendly thanks to natural refrigerant
  • OLED touch display with intuitive menu navigation
  • Good ratio between cooling capacity and size
  • USB-C and RS232 as standard, ethernet available as option
  • Illuminated, easy-to-
More Product Information