Nucleosides test mix, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
47310-U
47310-UEA 69.4 EUR
47310-U
Nucleosides test mix, analytical standard, Supelco®
Standaarden Standaarden voor chromatografie
Analytes:
Cytidine 50 μg/ml; Guanosine 25 μg/ml; Inosine 25 μg/ml; 1-Methyladenosine 25 μg/ml; 5-Methylcytidine 100 μg/ml; 2′-O-Methylcytidine 20 μg/ml; 3-Methylcytidine methosulfate 100 μg/ml; 7-Methylguanosine 25 μg/ml; 5-Methyluridine 50 μg/ml; β-Pseudouridine 25 μg/ml; 2-Thiocytidine dihydrate 10 μg/ml; Uridine 25 μg/ml