Methyleenchloride, anhydrous ≥99.7% (by GC) stabilised, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylene chloride

Stabilisatie: Stabilised with amylene
Packaged under argon.

47352.AK 47352.K2 47352.M1
47352.AKEA 42 EUR
47352.AK 47352.K2 47352.M1
Methyleenchloride, anhydrous ≥99.7% (by GC) stabilised, AcroSeal®
Methyleenchloride

Packaged under argon in resealable AcroSeal® bottles.

Formule: CH₂Cl₂
Kookpunt: 39…40 °C
Smeltpunt: –95 °C
Dichtheid: 1,325
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 14,6063
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.7% min
Water Content (Karl Fischer Titration) 0.01% max
Residue on Evaporation 3 ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR