Mycophenolic acid, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

475913-100 475913-500
475913-100EA 157 EUR
475913-100 475913-500
Mycophenolic acid, Millipore®
Mycophenolic acid
Formule: C₁₇H₂₀O₆
MW: 320.34 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036814
CAS nummer: 24280-93-1
EINECS: 246-119-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR