Dewarvaten, schaalvormig, type Sch-CAL
Leverancier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1191 1192 1193 1194 1195 1196 1197 1198 1673 1671 1675 1674 1672
478-0226EA 223 EUR
478-0226 478-4503 478-4505 478-4506 478-4507 478-4509 478-4510 478-4511 KGWA1673 KGWA1671 KGWA1675 KGWA1674 KGWA1672
Dewarvaten, schaalvormig, type Sch-CAL
Dewars en cryogene vaten
Silvered borosilicate glass, structured aluminium protective covering. For tempering of round bottom flasks, cooling of small samples. They are vacuum insulated and silvered.
  • Suitable as hot or cold bath
  • Can be used on a magnetic stirrer
  • The cover is anti-magnetic and stainless
Temperature range from −200 to +200 °C.
CAL flasks have an external stucco aluminium protective casing, alternatively the glass inserts (without casing) are also available.