Screw cap trays for 2D barcoded storage tubes
Leverancier: THERMO MATRIX TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4906 4477 4477BLU 4477GRE 4477GRY 4477PIN 4477PUR 4477RED 4477WHI 4477YEL
479-2066EA 81.5 EUR
479-2066 479-2052 479-2053 479-2054 479-2055 479-2056 479-2057 479-2058 479-2059 479-2060
Screw cap trays for 2D barcoded storage tubes
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Simplify sample storage with these color-coded caps and inserts.
Superior cap design
Gasketed caps ensure critical sample protection over the storage lifetime
Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
ScrewTop caps are compatible with storage down to −196 ºC
Cap options simplify sample identification
Cap trays feature standard microplate format compatible with automated equipment
Caps are available in seven color options to enableMore Product Information