Tissue vial box, blue, WHEATON® CryoFILE
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W651610-G W651610-Y W651610-P W651610-R W651610-W W651610-B
479-2112EA 203 EUR
479-2112 479-2113 479-2114 479-2117 479-2115 479-2116
Tissue vial box, blue, WHEATON® CryoFILE
Dozen Dozen voor vials
The CryoFILE tissue vial storage Box is used for batching and storing CryoELITE tissue vials.
CryoFILE partitions are numbered from 1 to 25 for easy content identification, with the numbering system printed on lid and bottom of box. CryoFILE tissue storage boxes are available in six colors, to provide easy sample identification. Made of water repellent material for longer durability, the boxes are able to withstand cryogenic temperatures to –196 °C.