VWR®, Cryoboxes, Half Size, Cardboard
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
479-7005EA 13.6 EUR
479-7005 479-2238 479-2239 479-2240 479-2241 479-2242
VWR®, Cryoboxes, Half Size, Cardboard
Dozen Dozen voor cryobuisjes
Cardboard, coated.
  • Waterproof coating
  • Freezable to –140 °C
  • Dividers available separately
  • Available in five colours
For storage of cryovials at low temperatures.
Order Now
Specificaties
Environmentally Preferable