Autoclaven, medium tot grote capaciteit, Priorclave
Leverancier: Priorclave
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EH100-SMART EH150-SMART EH200-SMART EH320-SMART EV100-SMART EV150-SMART EH200-BASE EV150-BASE EH100-BASE EH150-BASE EV100-BASE EH320-BASE
PRIOEH100-SMARTEA 0 EUR
PRIOEH100-SMART PRIOEH150-SMART PRIOEH200-SMART PRIOEH320-SMART PRIOEV100-SMART PRIOEV150-SMART 481-0915 481-0918 481-0913 481-0914 481-0917 481-0916
Autoclaven, medium tot grote capaciteit, Priorclave
Sterilisatoren Autoclaven (stoomsterilisatoren)
These versatile autoclaves are fully equipped for use in laboratories throughout the food, drink, dairy, health and pharmaceutical industries and offer a wide range of options and accessories to further improve their performance to suit individual load types. This flexibility is enhanced by the TACTROL™ microprocessor control system, specially developed for laboratory autoclaves, with progamming ranging from simple cycles to fully featured multi-program operation with logged records. All Priorclaves come complete with Biomaster antimicrobial coating, helping improved laboratory hygiene; itMore Product Information