β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate

4812-02EA 1426.39 EUR
4812-02 4812-04 4812-06 4812-08
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate

Suitable for use in biopharmaceutical manufacturing applications.

Formule: C₆H₁₃NO₄S·1H₂O
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149409
CAS nummer: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥98%
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Identification (by IR) Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Color of a 1M Alkaline Solution (Colorless) Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR