ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form), Calbiochem®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na

481971-1 481971-5 481971-500
481971-1EA 479 EUR
481971-1 481971-5 481971-500
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form), Calbiochem®
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
Formule: C₂₁H₂₇N₇NaO₁₇P₃
Smeltpunt: 175 to 178 °C
MDL Nummer: MFCD00036973
CAS nummer: 1184-16-3
EINECS: 214-664-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR