Nonidet® P 40 Substitute (NP-40), Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

492016-100 492016-1000 492016-500
492016-100EA 105 EUR
492016-100 492016-1000 492016-500
Nonidet® P 40 Substitute (NP-40), Millipore®
Nonidet® P 40 Substitute (NP-40)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00132851
CAS nummer: 9016-45-9
EINECS: 500-024-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR