Natriumchloride, ULTREX®, ultrapure, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Saline, NaCl, Sea salt, Halite

4924-05EA 607.11 EUR
4924-05
Natriumchloride, ULTREX®, ultrapure, J.T.Baker®
Natriumchloride

Certificate of lot analysis provided.

Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413
Smeltpunt: 801
Dichtheid: 2.16 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaCl) (dried basis), For Information Only
Loss on Drying at 650°C, For Information Only
Particulate Matter, For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only
Non-Metallic Impurities - Bromide (Br), For Information Only
Non-Metallic Impurities - Iodide (I), For Information Only
Non-Metallic Impurities - Nitrogen Compounds (as N), For Information Only
Non-Metallic Impurities - Phosphate (PO₄), For Information Only
Non-Metallic Impurities - Sulfur Compounds (as SO₄), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Lithium (Li), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Potassium (K), For Information Only
Metallic Impurities - Silver (Ag), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
Certificate Provided Reports Actual Lot Analysis

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR