N-Octyl-β-D-glucopyranoside, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

494460-1 494460-5
494460-1EA 385 EUR
494460-1 494460-5
N-Octyl-β-D-glucopyranoside, Millipore®
N-Octyl-β-D-glucopyranoside
Formule: C₁₄H₂₈O₆
MW: 292.37 g/mol
Smeltpunt: 110
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00063288
CAS nummer: 29836-26-8
EINECS: 249-887-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR