Cocarboxylase, EMPROVE® api suitable for use as an active pharmaceutical ingredient, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

5.00112.5000 5.00112.1000 5.00112.0100
5.00112.5000EA 0 EUR
5.00112.5000 5.00112.1000 5.00112.0100
Cocarboxylase, EMPROVE® api suitable for use as an active pharmaceutical ingredient, SAFC®
Cocarboxylase
Formule: C₁₂H₁₉ClN₄O₇P₂S
MDL Nummer: MFCD00038740
CAS nummer: 154-87-0
EINECS: 205-836-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR