Vitamin K1 (phytomenadione, phylloquinone) 1% lactose trituration

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

5.01758.5000
5.01758.5000EA 0 EUR
5.01758.5000
Vitamin K1 (phytomenadione, phylloquinone) 1% lactose trituration
Vitamin K1 (phytomenadione, phylloquinone)
Formule: C₃₁H₄₆O₂
MDL Nummer: MFCD00214063
CAS nummer: 84-80-0
EINECS: 201-564-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR