Discovery® Classical C18 HPLC Columns
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
504955 505129 569220-U 504971 59576-U 504947 50494721 50495521 505137 505161 505188 577507-U 59575-U 569222-U 569223-U 569224-U 569226-U 569229-U 569231-U
504955.EA 816 EUR
504955. 505129. 569220-U 504971. 59576-U 504947. 50494721. 50495521. 505137. 505161. 505188. 577507-U 59575-U 569222-U 569223-U 569224-U 569226-U 569229-U 569231-U
Discovery® Classical C18 HPLC Columns
Kolommen voor chromatografie
The Discovery® C18 columns provide the best suitable selectivity for many applications.
  • Excellent peak shape for all compounds
  • Very high stability
  • No-bleed properties for LC-MS applications
  • Impurity: <10 ppm metals
  • Surface coverage: 3 µmol/m²
Designed to meet the exacting requirements of pharmaceutical analysis and purification. Discovery® columns are also ideal for all application segments requiring reversed phase HPLC.
Bestelinformatie: Column test certificate available inside the box.