VWR®, Testzeven, 200×50 mm
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
510-0700EA 881 EUR
510-0700 510-0701 510-0702 510-0703 510-0704 510-0705 510-0706 510-0707 510-0708 510-0709 510-0710 510-0711 510-0712 510-0713 510-0714 510-0715 510-0716 510-0717 510-0718 510-0719 510-0720 510-0721 510-0722 510-0723 510-0724 510-0725 510-0726 510-0727 510-0728 510-0729 510-0730 510-0731 510-0732 510-0733 510-0734 510-0735 510-0736 510-0737 510-0738 510-0739 510-0740 510-0741 510-0742 510-0743 510-0744 510-0745 510-0746 510-0747 510-0748 510-0749 510-0750 510-0751 510-0752 510-0753 510-0754 510-0755 510-0756 510-0757 510-0758 510-0759 510-0760 510-0761 510-0762 510-0763 510-0764 510-0765 510-0766 510-0767 510-0768 510-0769 510-0770 510-0771 510-0772 510-0773 510-0774 510-0775 510-0776 510-0777 510-0778 510-0779 510-0780 510-0781 510-0782 510-0783 510-0784 510-0785 510-0786 510-0787 510-0788 510-0789 510-0790 510-0791 510-0792 510-0793 510-0794 510-0795 510-0796 510-0797 510-0798
VWR®, Testzeven, 200×50 mm
Zeven Testzeven
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to www.vwr.com for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.