Kwalitatief filterpapier, asvrije klassen, klasse 42, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1442-042 1442-047 1442-055 1442-070 1442-090 1442-110 1442-125 1442-150 1442-185 1442-240 1442-917 1442-320 1442-6551 1442-971
516-4301EA 20.1 EUR
516-4301 512-0593 512-0324 512-0325 512-0326 512-0327 512-0328 512-0329 512-0330 512-0009 512-0655 512-0627 512-0654 WHAT1442-971
Kwalitatief filterpapier, asvrije klassen, klasse 42, Whatman™
Filters Filterpapier Kwantitatief filterpapier
These filter papers made from high quality cotton linters have the finest retention of all ashless filter paper grades. Grade 42 is designed for gravimetric analysis and sample preparation for instrumental analysis.
  • Vaak gebruikt als een calciumcarbonaat filter
  • Alfacellulosegehalte ≥98%
These filter papers are designed for retention of extremely small particles. Other typical precipitates for gravimetric analysis include barium sulfate and metastannic acid for use in glazes, coatings and pigments. The high purity of these filters also makes themMore Product Information