Kwalitatief filterpapier, standaard klassen, klasse 3, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1003-055 1003-070 1003-090 1003-110 1003-125 1003-150 1003-185 1003-240 1003-320 1003-917 1003-323
512-1016EA 16.7 EUR
512-1016 512-1017 512-1018 512-1019 512-1020 512-1021 512-1022 512-0383 512-0464 512-1023 512-0465
Kwalitatief filterpapier, standaard klassen, klasse 3, Whatman™
Filters Filterpapier Kwalitatief filterpapier
A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
  • Thickness of 6 µm
Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Double the thickness of Grade 1 with still finer particle retention and excellent loading capacity; more precipitate can be held without clogging. The extra thickness gives increased wet strength and makes this grade highly suitable for use in BüchnerMore Product Information