Mixed cellulose ester membranes, sterile, 1,2 µm, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10408472
514-0946EA 264 EUR
514-0946
Mixed cellulose ester membranes, sterile, 1,2 µm, Whatman™
Filters Membraanfilters
Mixed cellulose ester, WME range, circles, gridded. Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high
More Product Information