MicroFunnel™ Plus filtertrechters
Leverancier: Cytiva (Formerly Pall Lab)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4807 4808 4809 4814 4823 4844 4807ME 4829ME
516-8964EA 612 EUR
516-8964 516-9890 516-9891 515-0052 515-0055 515-0157 516-1238 516-1239
MicroFunnel™ Plus filtertrechters
Trechters Membraanfiltertrechters
Ideal for monitoring purified water systems maintained at temperatures up to 90 °C (194 °F).
  • Multifunctional
  • Leakproof lid
  • Squeeze separation
  • Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0,2 and 0,45 µm Supor® (More Product Information
Order Now
Specificaties
Environmentally Preferable
Effectief filtratieoppervlak 13,46 cm² 13,46 cm² 13,46 cm²
Maximumtemperatuur Ambient 90 °C 90 °C
Afmetingen 76H×61W mm 76H×61W mm 89H×87W mm