Qualitative filter papers, technical, smooth, grade 3 hw
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FT-3-303-045 FT-4-303-070 FT-4-303-090 FT-4-303-500 FT2-303-580580 FT-3-303-060 FT-3-303-095 FT-3-303-120 FT-4-303-210 FT-3-303-055 FT-3-303-070 FT-3-303-090 FT-3-303-110 FT-3-303-125 FT-3-303-150 FT-3-303-185 FT-3-303-240 FT-3-303-270 FT-3-303-320 FT-3-303-385 FT-4-303-110 FT-4-303-125 FT-4-303-150 FT-4-303-185 FT-4-303-240 FT-4-303-270 FT-4-303-320 FT-4-303-385 FT-3-303-040 FT-3-303-050 FT-3-303-100 FT-3-303-085 FT-3-303-015 FT-4-303-290 FT-4-303-450 FT-3-303-080
515-1100EA 8.8 EUR
515-1100 515-1122 515-1123 515-1132 515-1616 SARTFT-3-303-060 SARTFT-3-303-095 SARTFT-3-303-120 SARTFT-4-303-210 515-1101 515-1102 515-1103 515-1104 515-1105 515-1106 515-1107 515-1108 515-1109 515-1110 515-1111 515-1124 515-1125 515-1126 515-1127 515-1128 515-1129 515-1130 515-1131 SARTFT-3-303-040 SARTFT-3-303-050 SARTFT-3-303-100 SARTFT-3-303-085 SARTFT-3-303-015 SARTFT-4-303-290 SARTFT-4-303-450 SARTFT-3-303-080
Qualitative filter papers, technical, smooth, grade 3 hw
Filters Filterpapier Kwalitatief filterpapier
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration. Used for essential oils; essences; salt solutions; tinctures; spirits; routine laboratory analyses.