Spuitfilters, Minisart® NML
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16532-GUK 16532-Q 16555-GUK 16592-K 17593-K 17593-Q 17594-K 17594-Q 17823-K 17823-Q 17824-K 17824-Q 17825-Q 17829-K 17829-Q 16534-K 16532-K 16592-Q 16553-K 16592-GUK 16541-K 16537-K 16533-Q 16533-K 16569-K 16533-GUK 16537-Q 16541-Q 16596-HNK 1751A-Q 1757A-Q 1659A-HYQ 16534-GUK 17597-K 17597-Q 16555-K 16555-Q 17598-K 17598-Q 16534-Q 17824-R S7594-FMOSK S6534-FMOSK S7597-FXOSK S6534-FMGUK S6534-FM-Q S7597-FX-Q S6555-FMOSK S7598-FXOSK S6555-FMGUK S6555-FM-Q S6596-FMOSK S6596-FM-Q S7598-FX-Q
516-0676EA 165 EUR
516-0676 512-3225 514-0672 512-3114 512-3116 512-3216 512-3118 512-3218 512-3106 512-3206 512-3120 512-3220 514-5002 512-3108 512-3208 514-7011 514-7010 512-3214 514-0152 516-0904 516-0680 516-0429 516-0901 516-0428 516-1160 516-0679 516-0902 516-0903 SART16596-HNK SART1751A-Q SART1757A-Q SART1659A-HYQ 611-0691 611-0707 611-0708 514-7013 514-7014 514-0674 514-0673 514-7012 SART17824-R 514-1287 514-1283 514-1288 514-1282 514-1284 514-1239 514-1285 514-1289 514-1235 514-1286 514-1237 514-1236 514-1290
Spuitfilters, Minisart® NML
Filters Spuitfilters
Minisart® NML with surfactant-free cellulose acetate (SFCA) is the best choice for all aqueous solutions and oils with a pH of 4 - 8. With a pore size of 0,2 µm that can be used for sterile filtration and has a male luer lock outlet. Only available in the EU/EEA and registered countries
  • Sterile and ready-to-use
  • SFCA in many pore sizes
  • Very low non-specific binding
  • PVC-free
  • Bidirectional use possible
Individually packed and sterile Minisart® NML syringe filters are optimal for immediate usage whenever sterile filtration or clarification and cellMore Product Information