SPE columns, PAH analysis, CHROMABOND® CN/SiOH
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
730135 730135G 730135.250
516-6010EA 297 EUR
516-6010 MANA730135G 516-0691
SPE columns, PAH analysis, CHROMABOND® CN/SiOH
Kolommen voor chromatografie
CHROMABOND® CN/SiOH is a combination phase for SPE analysis of PAHs.
  • Cyanopropyl phase for selective adsorption of polycyclic aromatics via π-π interactions
  • Unmodified silica phase for removal of polar impurities
Recommended for extraction of 16 PAHs according to EPA from soil samples as well as for an enrichment of PAHs in smoked meat products.
Order Now
Specificaties
Environmentally Preferable
pH-bereik 2 - 8