Kwalitatieve filterpapieren
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FT-3-206-055 FT-3-206-070 FT-3-206-090 FT-3-206-125 FT-3-206-185 FT-3-206-240 FT-3-207-125 FT-3-207-185 FT-3-207-240 FT-4-206-090 FT-4-206-110 FT-4-206-125 FT-4-206-150 FT-4-206-185 FT-4-206-270 FT-4-206-320 FT-4-206-385 FT-4-206-500 FT-4-207-110 FT-4-207-125 FT-4-207-150 FT-4-207-185 FT-4-207-270 FT-4-207-320 FT-4-207-385 FT2-207-580580 FT2-206-580580 FT-4-208-125 FT-4-208-110 FT-4-208-090 FT-4-207-090 FT-4-208-240 FT-4-209-320 FT-4-211-125 FT-4-210-320 FT-4-210-240 FT-4-210-185 FT-4-210-150 FT-4-210-125 FT-4-208-150 FT-4-209-385 FT-4-208-185 FT-4-209-270 FT-4-209-240 FT-4-209-185 FT-4-209-150 FT-4-209-125 FT-4-209-110 FT-4-208-385 FT-4-208-320 FT-4-208-270 FT-4-210-090 FT-3-209-110 FT-3-210-240 FT-3-210-185 FT-3-210-150 FT-3-210-125 FT-3-210-110 FT-3-210-090 FT-3-210-070 FT-3-210-055 FT-3-209-240 FT-3-209-185 FT-3-208-055 FT-3-209-125 FT-3-209-090 FT-3-209-070 FT-3-209-055 FT-3-208-240 FT-3-208-185 FT-3-208-150 FT-3-208-125 FT-3-208-110 FT-3-208-090 FT-3-209-150 FT-3-211-125 FT-3-211-150 FT-3-208-070 FT-3-207-090 FT-3-207-070 FT-3-207-055 FT-4-211-150 FT-3-211-185 FT-3-206-110 FT-3-206-150 FT-3-207-110 FT-4-207-240 FT-3-207-150 FT-4-211-185 FT-3-211-055 FT-3-211-070 FT-3-211-090 FT-3-211-110 FT2-208-580580 FT-4-211-090 FT-4-211-240 FT-3-207-045 FT-3-207-030 FT-3-207-340 FT-3-207-047 FT-3-207-018 FT-3-208-050 FT-3-206-040-5 FT-3-207-050 FT-3-209-006 FT2-211-580580 FT-3-207-060
515-1522EA 11.8 EUR
515-1522 515-1523 515-1524 515-1526 515-1528 515-1640 515-1641 515-1566 515-1567 515-1544 515-1545 515-1546 515-1547 515-1548 515-1550 515-1551 515-1552 515-1553 515-1585 515-1586 515-1587 515-1588 515-1647 515-1648 515-1649 515-1625 515-1624 512-0539 512-0538 512-0537 SARTFT-4-207-090 512-0542 SARTFT-4-209-320 SARTFT-4-211-125 SARTFT-4-210-320 512-0556 512-0555 512-0554 512-0553 512-0540 SARTFT-4-209-385 512-0541 512-0550 512-0549 512-0548 512-0547 512-0546 512-0545 SARTFT-4-208-385 SARTFT-4-208-320 512-0543 512-0551 516-7083 516-7095 516-7094 516-7093 516-7092 516-7091 516-7090 516-7089 516-7088 516-7087 516-7086 516-7072 516-7084 516-7082 516-7081 516-7080 516-7079 516-7078 516-7077 516-7076 516-7075 516-7074 516-7085 516-7063 516-7064 516-7073 516-7069 516-7068 516-7067 SARTFT-4-211-150 516-7065 515-1525 515-1527 515-1565 515-1589 515-1642 SARTFT-4-211-185 SARTFT-3-211-055 SARTFT-3-211-070 SARTFT-3-211-090 SARTFT-3-211-110 512-0531 SARTFT-4-211-090 SARTFT-4-211-240 SARTFT-3-207-045 SARTFT-3-207-030 SARTFT-3-207-340 SARTFT-3-207-047 SARTFT-3-207-018 SARTFT-3-208-050 SARTFT-3-206-040-5 SARTFT-3-207-050 SARTFT-3-209-006 SARTFT2-211-580580 SARTFT-3-207-060
Kwalitatieve filterpapieren
Filters Filterpapier Kwalitatief filterpapier
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration. They are made of refined pulp and linters with an >95% alpha-cellulose content, are very pure with an ash content <0,1%.
Grade 1288: For coarse-flocculent and voluminous precipitates, especially iron, aluminium and chromium hydroxides; copper, bismuth, cobalt and iron sulfides; various organic, colored metal deposits; determination of silicon in analysis of steelMore Product Information