Trechter volgens Dr. Hirsch
Leverancier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L126/00 L126/000 L126/4/0 L126/4/0A L126/5/0
519-2395EA 64.7 EUR
519-2395 519-2394 519-2392 519-2393 519-2391
Trechter volgens Dr. Hirsch
Trechters Filtertrechters Filtertrechters
Wit geglazuurd porselein.
Funnel with integral perforated filter plate can be used with paper or membrane discs.
Certificaten: DIN 12 905 (except 519-2392 and 519-2393).