Rotors for Centrifuges, 5810/5810 R
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5810709.008 5810711.002 5810718.007 5810725.003 5810743.001 5820710.004 5820740.000 5820754.001 5820755.008
521-0078EA 2090 EUR
521-0078 522-2730 521-0081 521-0145 522-0036 521-2072 521-1958 521-2074 521-2075
Rotors for Centrifuges, 5810/5810 R
Centrifuge Rotoren
Eppendorf Rotors for centrifuges 5810/5810 R - precision redefined. Tailor your high-capacity centrifugation with varied rotor options. Choose from a range of capacities and speeds, accessorize with versatile buckets, and ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering Renewable Energy Certificate, ISO 14001