Rotors for Centrifuges, 5910 Ri
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5895155.006 5895160.000 5895170.005 5895171.001 5895180.000 5895181.007
521-2324EA 1100 EUR
521-2324 521-2325 521-2326 521-2327 521-2328 521-2329
Rotors for Centrifuges, 5910 Ri
Centrifuge Rotoren
Eppendorf Rotors for Centrifuge 5910 Ri - precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options
More Product Information