Rotors for Centrifuge 5425/5425 R
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5495500.006 5495503.005 5495505.008 5495508.007 5495510.001 5495512.004
521-2481EA 750 EUR
521-2481 521-2482 521-2483 521-2484 521-2485 521-2486
Rotors for Centrifuge 5425/5425 R
Centrifuge Rotoren
Eppendorf rotors for centrifuge 5425/5425 R, precision meets versatility. Optimise your centrifuge with a range of capacities and speeds. Explore accessory options such as adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight and PTFE coated options available
More Product Information