Rotors for Centrifuges 5910 Ri and 5920 R
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5895120.008 5895121.004 5895130.003 5895135.005 5895150.004 5895200.001 5895201.008 5895175.007
521-2500EA 3320 EUR
521-2500 521-2501 521-2506 521-2507 521-2508 521-2332 521-2333 521-2689
Rotors for Centrifuges 5910 Ri and 5920 R
Centrifuge Rotoren
Eppendorf rotors for centrifuges 5910 Ri and 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
More Product Information