Microcentrifuges, 5425/5425 R (IVD-marked)
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5405000.212 5405000.611 5406000.011 5406000.216 5406000.410 5406000.615
521-2469EA 2500 EUR
521-2469 521-2477 521-3349 521-3353 521-3357 521-3361
Microcentrifuges, 5425/5425 R (IVD-marked)
Centrifuge Microcentrifuges
This air-cooled centrifuge covers a range of very frequent applications. Focusing on standard applications, it is an essential piece of equipment for modern labs ensuring optimal separation. Ideal for high-throughput users expecting a top level of ergonomic comfort as well as high flexibility with 6 rotor options.
  • QuickLock® technology for ergonomic rotor lid locking
  • OptiBowl® design for quiet operation, even without a rotor lid
  • Soft touch one-finger lid closure for ergonomic operation
  • Increased software features (10 acceleration and braking ramps, 3
More Product Information