Centrifuges, bench top, refrigerated, 5910Ri (IVD-marked)
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5943000.017 5943000.211 5943000.319 5943000.511
521-3383EA 12190 EUR
521-3383 521-3390 521-3395 521-3398
Centrifuges, bench top, refrigerated, 5910Ri (IVD-marked)
Centrifuge Tafelmodel centrifuges
The refrigerated benchtop centrifuge 5910 Ri stands out for its ease of use and versatility. With a range of innovative solutions in usability, documentation and user management. The centrifuge has designed to accommodate a particularly broad spectrum of applications. Cell harvests in bottles with capacities up to 1000 ml, large-scale DNA and RNA isolations as well as Ficoll® gradients for the purification of lymphocytes and monocytes are merely a few examples of the many areas of application.
  • 7" VisioNize® touchscreen interface with an intuitive user experience provides fast and error-
More Product Information